Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethyl alcohol, Pure, 64-17-5, MFCD00003568, 2 L
Catalog No. NC0410953
Encompass
Linear Formula: CH3CH2OH, Molecular Weight: 46.07, 200 proof, HPLC/spectrophotometric grade, Synonym: Ethanol, absolute alcohol, non-denatured ethanol. Pure ethyl alcohol (Ethanol) has been used to prepare: Ethanol solution for the synthesis of Ga-68 DOTATATE [DOTA-(Tyr3)-octreotate (where DOTA = 1,4,7,10-tetraazacyclo- dodecane-1,4,7,10-tetraacetic acid)] by an automated synthesis system. Aristolochic acid stock solution for the spectrophotometric analysis.
Catalog No. NC0410953 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 4598282L
Linear Formula: CH3CH2OH, Molecular Weight: 46.07, 200 proof, HPLC/spectrophotometric grade, Synonym: Ethanol, absolute alcohol, non-denatured ethanol. Pure ethyl alcohol (Ethanol) has been used to prepare: Ethanol solution for the synthesis of Ga-68 DOTATATE [DOTA-(Tyr3)-octreotate (where DOTA = 1,4,7,10-tetraazacyclo- dodecane-1,4,7,10-tetraacetic acid)] by an automated synthesis system. Aristolochic acid stock solution for the spectrophotometric analysis.