Learn More
Description
Specifications
Specifications
| Melting Point | -114°C |
| Color | Clear |
| Density | 6.62 |
| Quantity | 24 x 0.125 gal. |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Assay Percent Range | ≥99.5% |
| UN Number | UN1170 |
| Boiling Point | 78.3°C |
| Percent Purity | 100% |
| Show More |
Safety and Handling
| DOTInformation | DOT Class: 3, Packaging: II |
| Recommended Storage | Do not store alcohol above 120°F |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
