Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 200 proof, Molecular Biology Grade, Decon™ Labs
GSA_VAGREENER_CHOICE

Catalog No. 07678003 Shop All Decon Labs Products
Change view
Click to view available options
Quantity:
1 pt.
24 x 0.125 gal.
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
07-678-003 1 pt. Glass Bottle
07-678-002 24 x 0.125 gal. Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 07-678-003 Supplier Decon Laboratories, Inc. Supplier No. 3916EA

This product is not available for sale online. Please contact your sales rep or Customer Service at 1-800-766-7000.

missing translation for 'validMainframeMsg'

Pure Ethanol products are taxed according to the government schedule of excise taxes detailed in US CFR Part 27 (211.1119). This excise tax is included in the cost of this product. There is no need for Federal Permits

For the extraction and precipitation of DNA and RNA, can be used full strength or diluted with endonuclease-free water to desired dilution.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07 g/mol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name Ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Clear
Density 6.62
Quantity 1 pt.
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.5%
UN Number UN1170
Boiling Point 78.3°C
Percent Purity 100%
Grade Made to USP specifications, Tested for the presence of DNase and Rnase
Chemical Name or Material Ethanol, 200 Proof (100%)
Show More Show Less
DOTInformation DOT Class: 3, Packaging: II
Recommended Storage Do not store alcohol above 120°F
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.