Learn More
LAB ALLEY Lab Alley SDA 3C Ethanol 200 Proof (100%) Kosher Halal
SDP
Supplier:  LAB ALLEY EAS3C2001GAL
Encompass
This is a specially denatured alcohol formula 3C which contains the denaturant isopropyl alcohol at up to 5%. This formulation is compliant with United States Title 27 of the Code of Federal Regulations Section 21.151. Denaturants render denatured ethanol unfit for ingestion without otherwise interfering with ethanol chemistry. At Lab Alley we understand that high quality reagents are essential to the integrity of your laboratory and manufacturing projects. That is why we believe you deserve to have full confidence in the quality of your reagents. Lab Alley provides high quality reliable 200 proof SDA-3C ethanol ideal for your laboratory and manufacturing needs. 200 proof SDA 3C ethanol is in stock in all multiple sizes ranging from 500 ml to 55 gallons for quick shipping to you.
Catalog No. NC2468972
 Restricted Item