Learn More
Sigma Aldrich Fine Chemicals Biosciences ETHANOL 4L
Catalog No. NC2001176
Encompass
Ethanol gradient grade for liquid chromatography LiChrosolv 4L CAS#64-17-5 MFCD00003568 CH3CH2OH MW:46.07
Catalog No. NC2001176 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1117274000
Ethanol gradient grade for liquid chromatography LiChrosolv 4L CAS#64-17-5 MFCD00003568 CH3CH2OH MW:46.07