Learn More
IBI SCIENTIFIC 3P Ethanol | 64-17-5 | MFCD00003568 | 4 L
Catalog No. NC0753474
Encompass_Preferred
Ethanol (Alcohol, Anhydrous) - 4L Ethanol is widely used for precipitating nucleic acids. The nucleic acid precipitate, which is formed in the presence of moderate concentrations of monovalent cations, is recovered by centrifigution and redissolved.
Catalog No. NC0753474 Supplier IBI SCIENTIFIC 3P Supplier No. IB15724
Ethanol (Alcohol, Anhydrous) - 4L Ethanol is widely used for precipitating nucleic acids. The nucleic acid precipitate, which is formed in the presence of moderate concentrations of monovalent cations, is recovered by centrifigution and redissolved.