Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 70% v/v 140 Proof, Undenatured, Reagents

Catalog No. 01335389
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-389 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-389 Supplier Reagents Holdings Llc Supplier No. CE0032004A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethanol, 70% v/v 140 Proof, Undenatured, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym Ethyl Alcohol, Methylcarbinol
SMILES CCO

Specifications

Color Colorless
Quantity 4 L
Packaging Poly Bottle
Physical Form Liquid
Specific Gravity 0.8856
Boiling Point 78°C
IUPAC Name ethanol
Percent Purity 70%
Grade Reagent
Chemical Name or Material Ethanol, 70% v/v 140 Proof, Undenatured
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.