Learn More
Ethos Biosciences Ethanol 70PCT Denatured 5gal
Catalog No. 5000819199
Encompass_Preferred
Ethanol 70% Denatured is a solvent suitable for general laboratory pathology and research applications Commonly used as a dehydrating agent in tissue processing it is ideal for histology workflows slide preparation and reagent formulation This clear colorless alcohol provides consistent performance for reliable specimen handling and analytical results It is also widely used for cleaning solvent extraction instrument maintenance and other routine laboratory tasks Supplied in chemically compatible containers Ethanol 70% Denatured is appropriate for clinical research pharmaceutical and industrial laboratory environments
Catalog No. 50-008-19199 Supplier Ethos Biosciences Supplier No. 33375G
Ethanol 70% Denatured is a solvent suitable for general laboratory pathology and research applications Commonly used as a dehydrating agent in tissue processing it is ideal for histology workflows slide preparation and reagent formulation This clear colorless alcohol provides consistent performance for reliable specimen handling and analytical results It is also widely used for cleaning solvent extraction instrument maintenance and other routine laboratory tasks Supplied in chemically compatible containers Ethanol 70% Denatured is appropriate for clinical research pharmaceutical and industrial laboratory environments