Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

G-Biosciences Ethanol, 80%

Catalog No. S04207
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S04207 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04207 Supplier G-Biosciences Supplier No. 7862163
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Use as a wash solution for differential staining of Bacteria.

Use as a wash solution for differential staining of Bacteria.

Specifications

Quantity 250 mL
Percent Purity 80%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.