Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, 99.5%, ACS reagent, absolute, 200 proof
GSA_VA

Catalog No. AC615090040
Change view
Click to view available options
Quantity:
500 mL
1 L
2 L
4 L
Packaging:
HDPE Bottle
N/A
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC615090040 4 L HDPE Bottle
AC615095000 500 mL HDPE Bottle
AC615090010 1 L HDPE Bottle
AC615090020 2 L N/A
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC615090040 Supplier Thermo Scientific Chemicals Supplier No. 615090040
  • $884.50
    $514.65 / Each

    Save $369.85

    42% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $2,620.00
    $1,589.65 / Case of 4 EA

    Save $1,030.35

    39% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
 Restricted Item
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64-17-5 | C2H6O | 46.069 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Density 0.7900g/mL
Quantity 4 L
Infrared Spectrum Conforms to reference
Packaging HDPE Bottle
Refractive Index 1.3585 to 1.3605
Residue after Evaporation 0.001% max.
Assay Percent Range 99.5%
Titratable Acid 0.0005 meq/g max.
Linear Formula CH3CH2OH
Titratable Base 0.0002 meq/g max.
UN Number 1170
Water 0.2% max.
Merck Index 15, 3814
Boiling Point 78°C
Formula Weight 46.07
Percent Purity ≥99.5%
Grade ACS
Chemical Name or Material Ethanol, Absolute, 200 Proof
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.