Learn More
Sigma Aldrich Fine Chemicals Biosciences ETHANOL ABSOLUTE 200 PROOF 4L
Catalog No. NC1971050
Encompass
Ethanol absolute for analysis EMPARTA ACS, 99.5% (GC) CAS#64-17-5 MFCD00003568 CH3CH2OH MW:46.07
Catalog No. NC1971050 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1070174000
Ethanol absolute for analysis EMPARTA ACS, 99.5% (GC) CAS#64-17-5 MFCD00003568 CH3CH2OH MW:46.07