Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™
GSA_VA

Catalog No. BP28184 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP28184 4 L
BP2818100 100 mL
BP2818500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP28184
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request Bulk or Custom Formats

missing translation for 'validMainframeMsg'

Meets ACS Specifications

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Infrared Spectrum Conforms to reference
Physical Form Liquid
Residue after Evaporation 0.001% max.
Assay Percent Range Assay: ≥99.5% (v/v)
Titratable Acid 0.0005 mEq /g max.
Titratable Base 0.0002 mEq /g max.
Linear Formula C2H5OH
Water 0.2% max.
Merck Index 15, 3814
Boiling Point 78°C
Molecular Weight (g/mol) 46.07
Formula Weight 46.07
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Ethanol, Absolute (200 Proof)
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. May cause central nervous system effects. Aspiration hazard if swallowed - can enter lungs and cause damage. This substance has caused adverse reproductive and fetal effects in humans. Substances known to cause developmental toxicity in humans. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Obtain medical attention. Move to fresh air.

NFPA

Health:2
Flammability:3
Instability:0
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.