Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Solution III - Ethanol, ENG Scientific

Catalog No. ES7153
Change view
Click to view available options
Quantity:
2 x 250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES7153 2 x 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES7153 Supplier Eng Scientific Inc Supplier No. 7153
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Modified Toluidine Blue O Stain allows the Pneumocystis carinii cysts to be visualized more easily after staining. For Pneumocystis carinii Stain Kit (Modified Toluidine Blue O Stain).

  • Solution I - Toluidine Blue O Stain
  • Solution II - Ethanol (95%)
  • Solution III - Ethanol Solution IV - Xylene
  • Pneumocystis carinii is an opportunist which causes a diffuse interstitial pneumonia in patients with impaired immune systems.
  • The disease itself can be classified as epidemic or sporadic; the latter occurring in patients who have an underlying immunosuppression.
  • In this procedure, a sulfation reagent of glacial acetic acid and sulfuric acid (not included) is used for the removal of background material.
  • This allows the Pneumocystis carinii cysts to be visualized more easily after staining with Toluidine Blue O.
  • Diagnosis of Pneumocystis carinii pneumonia can frequently be made on bronchoalveolar lavage (BAL) or on touch preparations of pulmonary tissue (open lung biopsies and transbronchial biopsies).
  • Contents: Toluidine Blue O, HCl, Ethanol, Xylene
  • These solutions are made from certified dyes (when applicable).

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Quantity 2 x 250 mL
Packaging Bottle
Chemical Name or Material Ethanol

For in vitro diagnostic use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.