Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, anhydrous, denatured, 85.8%, 13.3% methanol, 0.9% MIBK

Catalog No. AA36642K2
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA36642K2 1 L
AA36642K7 4 L
AA36642M4 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA36642K2 Supplier Thermo Scientific Chemicals Supplier No. 36642K2
Add to Cart

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Denatured ethanol is acting as a solvent to remove the paint and ink stain on curtains, dress and upholstery. It is utilized as fuel in camping and primus stoves. It is likewise employed to clean the windows, manufacturing nail-polish and to test pectin levels in jam and jelly making.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • As a gasoline additive/substitute; octane booster
  • Very mobile liquid solvent
  • Used in lotions, tonics and perfumes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -100°C
Color Colorless
Density 0.811 g/mL
Quantity 1 L
Flash Point 11°C (52°F)
Odor Like alcohol
Physical Form Liquid
Refractive Index 1.361
Assay Percent Range 85.8%
Linear Formula CH3CH2OH
UN Number UN1993
Beilstein 1718733
Merck Index 14,3760
Boiling Point 80°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 46.08
Percent Purity 85.8%
Concentration or Composition (by Analyte or Components) 13.3% methanol, 0.9% MIBK
Chemical Name or Material Ethanol, Anhydrous, denatured
Show More Show Less
Hazard Category H225-H302-H319-H331-H370
Hazard Statement GHS H Statement
H225-H370
Highly flammable liquid and vapor.
Causes damage to organs.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P307+P311-P311-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 200-578-6
RTECSNumber KQ6300000
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.