Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol, anhydrous, denatured, HPLC Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA44134AE
Change view
Click to view available options
Quantity:
100 mL
Packaging:
ChemSeal™ Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AA44134AE 100 mL ChemSeal™ Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA44134AE Supplier Thermo Scientific Chemicals Supplier No. 044134AE
  • $70.60
    $57.65 / Each of 1

    Save $12.95

    18% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 64-17-5 | C2H6O | 46.069 g/mol

Ethanol, anhydrous, denatured is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -90°C
Color Colorless
Density 0.789 g/mL
Quantity 100 mL
Flash Point 11°C (52°F)
Odor Alcohol-like
Packaging ChemSeal™ Bottle
Physical Form Liquid
Refractive Index 1.361
Linear Formula CH3CH2OH
UN Number UN1987
Beilstein 1718733
Merck Index 14,3760
Boiling Point 78°C
Solubility Information Miscible with water and most organic solvents.
Formula Weight 46.08
Percent Purity 90%
Grade HPLC
Chemical Name or Material Ethanol, Anhydrous, denatured
Show More Show Less
Hazard Category H225-H302+H332-H319-H336-H371
Hazard Statement GHS H Statement
H225-H371
Highly flammable liquid and vapor.
May cause damage to organs.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P309+P311-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ALCOHOLS, N.O.S.
EINECSNumber 200-578-6
RTECSNumber KQ6300000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.