Learn More
Bioworld ETHANOL ANHYDROUS DENATURED
Catalog No. 50255963
Encompass_Preferred
This item has a minimum order quantity of 3 per supplier requirements. Biotechnology Grade Specifications: Identification (IR)(P/F)- Pass Methanol(%)- =5.0 Moisture (KF)(%) - =1.0 Purity(%)- 94.9-96.0 Specific Gravity- =0.7964
Catalog No. 50-255-963 Supplier Bioworld Supplier No. 405700001
This item has a minimum order quantity of 3 per supplier requirements. Biotechnology Grade Specifications: Identification (IR)(P/F)- Pass Methanol(%)- =5.0 Moisture (KF)(%) - =1.0 Purity(%)- 94.9-96.0 Specific Gravity- =0.7964