Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol 'C' Solvent/Water, 95/5 (w/w), Reagents

Catalog No. 01335370
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-370 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-370 Supplier Reagents Holdings Llc Supplier No. CE2410004C
 Restricted Item
Add to Cart
Add to Cart

Ethanol CM Solvent/Water, 95/5 (w/w), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ethanol C Solvent/Water, 95/5 (w/w)
CAS 64-17-5, 7732-18-5, 67-56-1
Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.808
Molecular Formula CH3CH2OH
Synonym Ethyl Alcohol, Methylcarbinol
Boiling Point 79°C
Percent Purity 95%
Grade Reagent
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.