Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol/Cyclohexane 50/50 v/v Made with HPLC grade materials, Reagents

Catalog No. 01335391
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-391 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-391 Supplier Reagents Holdings Llc Supplier No. CE0027004C
Add to Cart
Add to Cart

Ethanol/Cyclohexane 50/50 v/v Made with HPLC grade materials, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 110-82-7 , 64-17-5 , 67-63-0
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifications

Color Colorless
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.7844
Boiling Point 72°C
Grade Reagent
Chemical Name or Material Ethanol/Cyclohexane 50/50 v/v Made with HPLC grade materials
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.