Learn More
Medchemexpress LLC Ethanol-d6 | 1516-08-1 | 99.3% | 52.11 | C2D6O | 500 MG
SDP
Catalog No. 5000208784
Encompass_Preferred
Ethanol-d6 is the deuterium-labeled form of ethanol used as an analytical and research solvent for NMR spectroscopy and isotope-tracing studies. It is supplied as a colorless to light yellow liquid with a reported purity of 99.32% and a molecular weight of 52.11 g/mol, and it has the formula C2D6O.
  • Deuterium-labeled solvent suitable for NMR and tracer experiments.
  • High stated purity (99.3%) for reliable analytical results.
  • Low-viscosity liquid physical state for easy handling and dilution.
  • Stable storage recommendations for long-term integrity.
  • Available in multiple pack sizes to match experimental scale.
Catalog No. 50-002-08784 Supplier Medchemexpress LLC Supplier No. HYW091739500MG
Ethanol-d6 is the deuterium-labeled form of ethanol used as an analytical and research solvent for NMR spectroscopy and isotope-tracing studies. It is supplied as a colorless to light yellow liquid with a reported purity of 99.32% and a molecular weight of 52.11 g/mol, and it has the formula C2D6O.
  • Deuterium-labeled solvent suitable for NMR and tracer experiments.
  • High stated purity (99.3%) for reliable analytical results.
  • Low-viscosity liquid physical state for easy handling and dilution.
  • Stable storage recommendations for long-term integrity.
  • Available in multiple pack sizes to match experimental scale.