Learn More
Sigma Aldrich Fine Chemicals Biosciences Ethanol d6 anhydrous 99.55G
Catalog No. 502898005
Encompass
Ethanol-d6 is a deuterated derivative of ethanol. It has an isotopic purity of 99.5atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical).
Catalog No. 50-289-8005 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 1864145G
Ethanol-d6 is a deuterated derivative of ethanol. It has an isotopic purity of 99.5atom%D. It is a standard purity solvent suitable for routine NMR analyses (conducted at ambient temperatures where quality is less critical).