Learn More
Bioworld Ethanol, Denatured, 72%, 4 L
Catalog No. 50255971
Encompass_Preferred
It is a volatile, flammable, colorless straight-chain, primary alcohol and used as solvent and fuel.Appearance: Colourless liquidAtomic Number: Not available.Boiling Point : 78°CChEBI : Not available.Crystal_Structure : Not available.Density : 0.789 g/cm3Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.IUPAC_Name: EthanolMelting Point: W22;114°CMolar Mass: 46.07 g/molNFPA 704: Not available.RTECS Number: KQ6300000Synonyms: Ethyl Alcohol, Reagent Alcohol
Catalog No. 50-255-971 Supplier Bioworld Supplier No. 405700123
It is a volatile, flammable, colorless straight-chain, primary alcohol and used as solvent and fuel.Appearance: Colourless liquidAtomic Number: Not available.Boiling Point : 78°CChEBI : Not available.Crystal_Structure : Not available.Density : 0.789 g/cm3Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.IUPAC_Name: EthanolMelting Point: W22;114°CMolar Mass: 46.07 g/molNFPA 704: Not available.RTECS Number: KQ6300000Synonyms: Ethyl Alcohol, Reagent Alcohol