Learn More
Bioworld Ethanol, Denatured, 63%, 500 mL
Catalog No. 50255972
Encompass_Preferred
Denatured with 2.9-3.5% methanol and 3.2-3.9% 2-propanol. Suitable for cytology and histology.Appearance: Clear, colorless liquid.Atomic Number: Not available.Boiling Point : 78°CChEBI : Not available.Crystal_Structure : Not available.Density : 0.789 g/cm3Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.IUPAC_Name: EthanolMelting Point: W22;114°CMolar Mass: 46.07g/molNFPA 704: Not available.RTECS Number: KQ6300000Synonyms: Ethyl Alcohol; Reagent Alcohol
Catalog No. 50-255-972 Supplier Bioworld Supplier No. 405700131
Denatured with 2.9-3.5% methanol and 3.2-3.9% 2-propanol. Suitable for cytology and histology.Appearance: Clear, colorless liquid.Atomic Number: Not available.Boiling Point : 78°CChEBI : Not available.Crystal_Structure : Not available.Density : 0.789 g/cm3Electron Configuration : Not available.InChI : Not available.InChIKey : Not available.Ionization Energy : Not available.IUPAC_Name: EthanolMelting Point: W22;114°CMolar Mass: 46.07g/molNFPA 704: Not available.RTECS Number: KQ6300000Synonyms: Ethyl Alcohol; Reagent Alcohol