Learn More
Supply Solutions MSI Brand - Ethanol Industry HPLC Standard. Size: 5x10ml; Grade: HPLC; ppm: 12%.; Size: 5x10ml; Grade: HPLC; ppm: 0.12; CAS: ; Hazardous: Y; Container Material: Glass Vial
Catalog No. NC0940787
Encompass
A key measurement is the residual sugar and ethanol concentrations in the fermentation broth. Fuel ethanol facilities use High-Performance Liquid Chromatography (HPLC) as the technique of choice to monitor the ethanol fermentation process. HPLC permits detailed monitoring of the complete cycle, including conversion of the sugars to ethanol and ethanol breakdown to acetic acid.; Size: 5x10ml; Grade: HPLC; ppm: 0.12; CAS: ; Hazardous: Y; Container Material: Glass Vial; Contains varying amounts: Ethanol, DP4, DP3, Maltose, Glucose, Lactic Acid, Acetic Acid, Glycerol; Check standard methodology is a tool for collecting data on the measurement process to expose errors that afflict the process over time.; Shelf Life: 12 Months
Catalog No. NC0940787 Supplier Supply Solutions Supplier No. MSI HPLC1000.1
A key measurement is the residual sugar and ethanol concentrations in the fermentation broth. Fuel ethanol facilities use High-Performance Liquid Chromatography (HPLC) as the technique of choice to monitor the ethanol fermentation process. HPLC permits detailed monitoring of the complete cycle, including conversion of the sugars to ethanol and ethanol breakdown to acetic acid.; Size: 5x10ml; Grade: HPLC; ppm: 0.12; CAS: ; Hazardous: Y; Container Material: Glass Vial; Contains varying amounts: Ethanol, DP4, DP3, Maltose, Glucose, Lactic Acid, Acetic Acid, Glycerol; Check standard methodology is a tool for collecting data on the measurement process to expose errors that afflict the process over time.; Shelf Life: 12 Months