Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol Solution 70%, Molecular Biology Grade, Fisher BioReagents™

Catalog No. BP82014 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BP82014 4 L
BP82011 1 L
BP8201500 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. BP82014
  • $518.50
    $275.65 / Each

    Save $242.85

    47% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
  • $1,154.00
    $613.65 / Case of 4 EA

    Save $540.35

    47% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Use to concentrate nucleic acid samples by precipitation

  • DNase, and Protease tested to verify absence of hydrolytic enzymes
  • Combine this blend with commercial “mini-prep“ kits or traditional extraction
  • ACS grade Absolute Ethanol used in preparation of solution
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5 , 7732-18-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Colorless
CAS Min % 70%
CAS Max % 30%
Density 0.80g/cm3
Quantity 4 L
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation ≤0.001%
Water 28%-32% (GC-TCD)
Boiling Point 78°C
Grade Molecular Biology
Chemical Name or Material 70% Ethanol Solution
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.