Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol Solution, MilliporeSigma™

Catalog No. 50947987 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-947-987 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-947-987 Supplier MilliporeSigma Supplier No. EX02811
Add to Cart
Add to Cart

Chemical Identifiers

CAS 64-17-5

Specifications

Quantity 4 L
Flash Point 10.6°C
Packaging Glass Bottle (Pre-packed)
Recommended Storage Store at room temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.