Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanol Standards 10 (v/v) 10 (v/v) in H2O, MilliporeSigma™ Supelco™

Catalog No. 111018716 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8716 10 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8716 Supplier MilliporeSigma Supelco Supplier No. E238510AMP
Add to Cart
Add to Cart

The product is a 10% (v/v) solution of ethanol in water. Ethanol, a primary alcohol, is an important raw material for the synthesis of various compounds. The impact of ethanol/water fumigation on the performance of diesel engines has been studied.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 64-17-5
Flash Point 44°C
Concentration 10% (v/v) in H 2 O
Linear Formula C2H5 OH
MDL Number MFCD00003568
Synonym Ethanol solution; Ethyl alcohol
Type Analytical Standard
Quantity 10 x 1 mL
Formula Weight 46.07
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.