Learn More
YSI 3P LifeScience ETHANOL STANDARDS KIT
Catalog No. 50200133
Encompass
2.0 g/L and 3.2 g/L Ethanol Standard solutions for use with YSI 2700, 2900 Series and 7100 Analyzers. Kit contains 3 bottles of 2.0 g/L standard and 2 bottles of 3.2 g/L standard.
Catalog No. 50-200-133 Supplier YSI 3P LifeScience Supplier No. 2790
2.0 g/L and 3.2 g/L Ethanol Standard solutions for use with YSI 2700, 2900 Series and 7100 Analyzers. Kit contains 3 bottles of 2.0 g/L standard and 2 bottles of 3.2 g/L standard.