Learn More
FUJIFILM Biosciences Ethanol Super Dehydrated 99.5
Catalog No. 5000450268
Encompass_Preferred
Used for organic synthesis and research A solvent of super dehydrated grade for organic synthesis that contains not more than 10 ppm 0 001% of water The special cap employed in 100-mL and 500-mL containers facilitate aspiration of the solvent using a syringe without opening the container Filled up with inactive gas Highly flammable and needs handling with precautions For research use only RUO
Catalog No. 50-004-50268 Supplier FUJIFILM Biosciences Supplier No. 05008425
Used for organic synthesis and research A solvent of super dehydrated grade for organic synthesis that contains not more than 10 ppm 0 001% of water The special cap employed in 100-mL and 500-mL containers facilitate aspiration of the solvent using a syringe without opening the container Filled up with inactive gas Highly flammable and needs handling with precautions For research use only RUO