Learn More
Medchemexpress LLC Ethanolamine-13C2 hydrochloride | 1173019-25-4 | 98.0% | 99.53 | 13C2H8ClNO | 10 MG
SDP
Catalog No. 5000300078
Encompass_Preferred
Ethanolamine-13C2 hydrochloride is the 13C-labeled form of ethanolamine hydrochloride supplied as a hydrochloride salt. It is used as an isotope-labeled reagent and internal standard in biochemical, metabolomics, and mass spectrometry workflows.
  • Isotopically labeled with two 13C atoms.
  • Hydrochloride salt, supplied as a solid.
  • High purity: 98.0%.
  • Molecular weight: 99.53 g/mol.
  • Suitable as an internal standard for mass spectrometry and metabolomics.
  • Available in small pack sizes (1 mg, 5 mg, 10 mg).
Catalog No. 50-003-00078 Supplier Medchemexpress LLC Supplier No. HYW035903S10MG
Ethanolamine-13C2 hydrochloride is the 13C-labeled form of ethanolamine hydrochloride supplied as a hydrochloride salt. It is used as an isotope-labeled reagent and internal standard in biochemical, metabolomics, and mass spectrometry workflows.
  • Isotopically labeled with two 13C atoms.
  • Hydrochloride salt, supplied as a solid.
  • High purity: 98.0%.
  • Molecular weight: 99.53 g/mol.
  • Suitable as an internal standard for mass spectrometry and metabolomics.
  • Available in small pack sizes (1 mg, 5 mg, 10 mg).