Learn More
Medchemexpress LLC Ethanolamine-13C2 hydrochloride | 1173019-25-4 | MFCD00144263 | 98.0% | 99.53 g/mol | 13C2H8ClNO | 5 MG
SDP
Catalog No. 5000301551
Encompass_Preferred
Ethanolamine-13C2 hydrochloride is a stable-isotope labeled form of ethanolamine (1,2-13C2) supplied as the hydrochloride salt. It is intended for use as an internal standard or tracer in mass spectrometry, metabolomics, and biochemical assay development.
  • Stable 13C2 isotopic labeling for accurate tracing and quantification.
  • High purity suitable for analytical and mass spectrometry applications.
  • Provided as the hydrochloride salt for easier handling and solubility.
  • Available in small pack sizes appropriate for standards and method development.
  • Product documentation includes data sheet, certificate of analysis, and SDS.
Catalog No. 50-003-01551 Supplier Medchemexpress LLC Supplier No. HYW035903S5MG
Add to Cart
Add to Cart
Ethanolamine-13C2 hydrochloride is a stable-isotope labeled form of ethanolamine (1,2-13C2) supplied as the hydrochloride salt. It is intended for use as an internal standard or tracer in mass spectrometry, metabolomics, and biochemical assay development.
  • Stable 13C2 isotopic labeling for accurate tracing and quantification.
  • High purity suitable for analytical and mass spectrometry applications.
  • Provided as the hydrochloride salt for easier handling and solubility.
  • Available in small pack sizes appropriate for standards and method development.
  • Product documentation includes data sheet, certificate of analysis, and SDS.