Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, 20% w/v, Reagents

Catalog No. 01335409
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-335-409 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-335-409 Supplier Reagents Holdings Llc Supplier No. CE00010010E2

missing translation for 'validMainframeMsg'

Ethanolamine, 20% w/v, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

CAS 7732-18-5, 141-43-5
Color Colorless
Boiling Point 100°C
Packaging HDPE Winpak
Molecular Formula H2O
Quantity 10 L
Specific Gravity 1.0024
Synonym 2-Aminoethanol, Monoethanolamine
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
SMILES O
IUPAC Name water
Molecular Weight (g/mol) 18.02
Percent Purity 20%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Ethanolamine, 20% w/v
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.