Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Ethanolamine

Catalog No. 1501425L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
2.5 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-014-25L 2.5 L
15-014-1L 1 L
15-014-100ML 100 mL
15-014-25ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 15-014-25L Supplier Sigma Aldrich Supplier No. 150142.5L
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Ethanolamine

Specifications

CAS 141-43-5
Melting Point 10°C to 11°C
Density 1.012 g/mL at 25°C
Boiling Point 170°C (69°C to 70°C (10 mmHg)
Molecular Formula C2H7NO
Linear Formula NH2CH2CH2OH
Refractive Index n20/D 1.454
MDL Number MFCD00008183
Quantity 2.5 L
Synonym 2-Aminoethanol; 2-Aminoethyl alcohol; ETA; MEA; MEA 90; MEA-LCI; Monoethanolamine
Molecular Weight (g/mol) 61.08 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.