Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019104 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9104 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9104 Supplier MilliporeSigma Supelco Supplier No. 002785MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ethanolamine is a colourless organic compound. It is a strong base and is miscible in water, alcohol, carbon tetrachloride and chloroform. It is highly toxic in nature and is a strong irritant. It is derived from ethylene oxide reacting with ammonia forming mono-, di-, and triethanolamine. It is widely used in manufacturing detergents when reacting with fatty acid. It is most commonly used a corrosion inhibitor. Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 141-43-5
Ignition Point 1436°F
Vapor Pressure 0.2 mmHg (20°C)
Vapor Density 2.1 (vs. air)
Density 1.012 g/mL (at 25°C)
Flash Point 91°C - Pensky-Martens closed cup
Linear Formula NH2 CH2 CH2 OH
MDL Number MFCD00008183
Melting Point 10°C to 11°C
Boiling Point 170°C; 69°C to 70°C (10 mmHg)
Synonym 2-Aminoethanol; 2-Aminoethyl alcohol; ETA; MEA; MEA 90; MEA-LCI; Monoethanolamine
Type Analytical Standard
Quantity 5 mL
Formula Weight 61.08
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.