Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethanolamine benzoate hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog No. p-7028512
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2586414 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2586414 Supplier Thermo Scientific Chemicals Supplier No. H2586414
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 545375-30-2
Molecular Formula C9H12ClNO2
Molecular Weight (g/mol) 201.65
MDL Number MFCD09953460
InChI Key ZXTMLOSVFWKYCL-UHFFFAOYSA-N
Synonym ethanolamine benzoate hydrochloride, 2-aminoethyl benzoate hydrochloride, ethanolamine benzoate hydrochloride, 98+%, 2-benzoyloxyethylamine hydrochloride
PubChem CID 67778770
IUPAC Name 2-aminoethyl benzoate;hydrochloride
SMILES Cl.NCCOC(=O)C1=CC=CC=C1

Specifications

Melting Point 141°C to 142°C
Quantity 25 g
Formula Weight 201.65
Percent Purity ≥98%
Chemical Name or Material Ethanolamine benzoate hydrochloride
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.