Learn More
Medchemexpress LLC Ethanolamine hydrochloride | 2002-24-6 | MFCD00012892 | >99.0% | 97.54 g/mol | C2H8ClNO | 50 G
SDP
Catalog No. 5000299442
Encompass_Preferred
Ethanolamine hydrochloride is the hydrochloride salt of ethanolamine, provided as a high-purity biochemical reagent for use as a buffer component and pH modifier in life-science research and analytical workflows. It is commonly used in biochemical assays, cell culture buffers, and reagent preparations, and is supplied as a stable solid for laboratory storage.
  • High purity (≥99.0%) suitable for analytical and biochemical applications.
  • White to off-white solid for easy handling and storage.
  • Acts as a buffer component and pH modifier in biological assays.
  • Compatible with cell culture and reagent preparations.
  • Store sealed, away from moisture, at recommended temperatures for stability.
Catalog No. 50-002-99442 Supplier Medchemexpress LLC Supplier No. HYW03590350G
Ethanolamine hydrochloride is the hydrochloride salt of ethanolamine, provided as a high-purity biochemical reagent for use as a buffer component and pH modifier in life-science research and analytical workflows. It is commonly used in biochemical assays, cell culture buffers, and reagent preparations, and is supplied as a stable solid for laboratory storage.
  • High purity (≥99.0%) suitable for analytical and biochemical applications.
  • White to off-white solid for easy handling and storage.
  • Acts as a buffer component and pH modifier in biological assays.
  • Compatible with cell culture and reagent preparations.
  • Store sealed, away from moisture, at recommended temperatures for stability.