Learn More
Selleck Chemical LLC Ethanolamine hydrochloride
SDP
Catalog No. 5000003346
Encompass_Preferred
Ethanolamine (2-Aminoethanol) is a viscous hygroscopic amino alcohol with an ammoniacal odor It is widely distributed in biological tissue and is a component of lecithin
Catalog No. 50-000-03346 Supplier Selleck Chemical LLC Supplier No. S62101G
Ethanolamine (2-Aminoethanol) is a viscous hygroscopic amino alcohol with an ammoniacal odor It is widely distributed in biological tissue and is a component of lecithin