Learn More
Medchemexpress LLC Ethanone, 1-(2,5-dihydroxyphenyl)- | 490-78-8 | 152.15 | 1 G
SDP
Catalog No. 5000447856
Encompass_Preferred
2,5-Dihydroxyacetophenone is isolated from Rehmannia glutinosa and functions by inhibiting the production of inflammatory mediators in activated macrophages. It primarily achieves this by blocking the ERK1/2 and NF-κB signaling pathways. This compound is intended for research use only and is not suitable for patient applications.
  • Inhibits NO production through suppression of iNOS expression
  • Decreases levels of pro-inflammatory cytokines such as TNF-α and IL-6
  • Blocks ERK1/2 and NF-κB signaling pathways
  • Potently inhibits the phosphorylation of extracellular signal-related kinase (ERK) 1/2
  • Potently inhibits the nuclear translocation of nuclear factor-κB (NF-κB) p65
Catalog No. 50-004-47856 Supplier Medchemexpress LLC Supplier No. HYW0011741G
2,5-Dihydroxyacetophenone is isolated from Rehmannia glutinosa and functions by inhibiting the production of inflammatory mediators in activated macrophages. It primarily achieves this by blocking the ERK1/2 and NF-κB signaling pathways. This compound is intended for research use only and is not suitable for patient applications.
  • Inhibits NO production through suppression of iNOS expression
  • Decreases levels of pro-inflammatory cytokines such as TNF-α and IL-6
  • Blocks ERK1/2 and NF-κB signaling pathways
  • Potently inhibits the phosphorylation of extracellular signal-related kinase (ERK) 1/2
  • Potently inhibits the nuclear translocation of nuclear factor-κB (NF-κB) p65