Learn More
Medchemexpress LLC Ethanone, 1,2-diphenyl- | 451-40-1 | MFCD00003081 | 25g
SDP
Catalog No. 5000266233
Encompass_Preferred
2-Phenylacetophenone has broad-spectrum efflux pump inhibition activity 2-Phenylacetophenone is a benzoin derivative used as a photoinitiator in vinyl polymerization[1]
Catalog No. 50-002-66233 Supplier Medchemexpress LLC Supplier No. HYY177725G
2-Phenylacetophenone has broad-spectrum efflux pump inhibition activity 2-Phenylacetophenone is a benzoin derivative used as a photoinitiator in vinyl polymerization[1]