Learn More
Medchemexpress LLC Ethanone, 1-(2-furanyl)- | 1192-62-7 | MFCD00003242 | 100.0% | 110.11 g/mol | C6H6O2 | 100 G
SDP
Catalog No. 5000300986
Encompass_Preferred
2-Acetylfuran is a low-molecular-weight furan derivative used as a flavor compound and as an intermediate in organic synthesis. It exhibits a sweet, caramel-like aroma and is commonly used in flavor and fragrance formulation, chemical research, and as a standard in analytical methods. High-purity material and supporting documentation are available for laboratory and industrial use.
  • High purity suitable for research and synthesis
  • Distinct caramel-like aroma for flavor formulation
  • Versatile intermediate for organic synthesis
  • Available in multiple package sizes for scale flexibility
  • Documented safety and quality data available
Catalog No. 50-003-00986 Supplier Medchemexpress LLC Supplier No. HYW015912100G
2-Acetylfuran is a low-molecular-weight furan derivative used as a flavor compound and as an intermediate in organic synthesis. It exhibits a sweet, caramel-like aroma and is commonly used in flavor and fragrance formulation, chemical research, and as a standard in analytical methods. High-purity material and supporting documentation are available for laboratory and industrial use.
  • High purity suitable for research and synthesis
  • Distinct caramel-like aroma for flavor formulation
  • Versatile intermediate for organic synthesis
  • Available in multiple package sizes for scale flexibility
  • Documented safety and quality data available