Learn More
Medchemexpress LLC Ethanone, 1-(2-pyridinyl)-, 2-(6-methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone | 327031-55-0 | MFCD01305422 | 99.8% | 317.35 g/mol | C17H15N7 | 10MG
SDP
Catalog No. 5000235143
Encompass_Preferred
VLX600 is an iron-chelating inhibitor of oxidative phosphorylation used as a research tool to induce mitochondrial dysfunction, shift cellular metabolism toward glycolysis, and trigger autophagy and selective cytotoxicity in malignant cells. It is supplied for research use and is characterized by CAS 327031-55-0, molecular formula C17H15N7, and molecular weight 317.35 g/mol.
  • Iron-chelating inhibitor of oxidative phosphorylation that disrupts mitochondrial function.
  • Induces a metabolic shift toward glycolysis in treated cells.
  • Promotes autophagy and selective cytotoxicity in malignant or quiescent cancer cells.
  • High chemical purity, typically 99.8%.
  • Supplied in small pack sizes suitable for in vitro research (for example, 10 mg).
Catalog No. 50-002-35143 Supplier Medchemexpress LLC Supplier No. HY1240610MG
VLX600 is an iron-chelating inhibitor of oxidative phosphorylation used as a research tool to induce mitochondrial dysfunction, shift cellular metabolism toward glycolysis, and trigger autophagy and selective cytotoxicity in malignant cells. It is supplied for research use and is characterized by CAS 327031-55-0, molecular formula C17H15N7, and molecular weight 317.35 g/mol.
  • Iron-chelating inhibitor of oxidative phosphorylation that disrupts mitochondrial function.
  • Induces a metabolic shift toward glycolysis in treated cells.
  • Promotes autophagy and selective cytotoxicity in malignant or quiescent cancer cells.
  • High chemical purity, typically 99.8%.
  • Supplied in small pack sizes suitable for in vitro research (for example, 10 mg).