Learn More
Medchemexpress LLC Ethanone, 1-(2-pyridinyl)-, 2-(6-methyl-5H-1,2,4-triazino[5,6-b]indol-3-yl)hydrazone | 327031-55-0 | MFCD01305422 | 99.8% | 317.35 g/mol | C17H15N7 | 50MG
SDP
Catalog No. 5000237387
Encompass_Preferred
VLX600 is a small-molecule iron-chelating inhibitor of oxidative phosphorylation that disrupts mitochondrial function, shifts metabolism toward glycolysis, and exhibits selective cytotoxicity against malignant and quiescent cancer cells. It is supplied as a research-grade powder for preclinical biochemical and cellular studies; follow recommended storage and handling.
  • Iron-chelating inhibitor of oxidative phosphorylation and modulator of mitochondrial metabolism.
  • Selective cytotoxicity against quiescent and malignant cancer cells in preclinical studies.
  • High purity (99.8%) appropriate for research-grade assays.
  • Supplied as a powder; recommended cold storage to preserve stability.
  • Chemical formula C17H15N7 and molecular weight 317.35 g/mol.
Catalog No. 50-002-37387 Supplier Medchemexpress LLC Supplier No. HY1240650MG
VLX600 is a small-molecule iron-chelating inhibitor of oxidative phosphorylation that disrupts mitochondrial function, shifts metabolism toward glycolysis, and exhibits selective cytotoxicity against malignant and quiescent cancer cells. It is supplied as a research-grade powder for preclinical biochemical and cellular studies; follow recommended storage and handling.
  • Iron-chelating inhibitor of oxidative phosphorylation and modulator of mitochondrial metabolism.
  • Selective cytotoxicity against quiescent and malignant cancer cells in preclinical studies.
  • High purity (99.8%) appropriate for research-grade assays.
  • Supplied as a powder; recommended cold storage to preserve stability.
  • Chemical formula C17H15N7 and molecular weight 317.35 g/mol.