Learn More
Medchemexpress LLC Ethanone, 1-[(3S)-3-amino-1-pyrrolidinyl]-2-(3,5-difluoro-2-PhO)cyclopropylamino HCl | 2262489-89-2 | 98.1% | C22H26ClF2N3O2 | 10MG
SDP
Catalog No. 5000259240
Encompass_Preferred
S2116 is an N-alkylated tranylcypromine derivative and potent lysine-specific demethylase 1 (LSD1) inhibitor used in epigenetic research. It is supplied as a solid or as a DMSO solution for biochemical and cellular studies; refer to the certificate of analysis for batch-specific purity and handling information.
  • Potent inhibitor of lysine-specific demethylase 1 (LSD1).
  • High purity, 98.1% by HPLC.
  • Available as solid powder and 10 mM solution in DMSO.
  • Chemical formula C22H26ClF2N3O2 and molecular weight 437.91 g/mol.
  • Intended for research use only; not for human use.
Catalog No. 50-002-59240 Supplier Medchemexpress LLC Supplier No. HY13652210MG
S2116 is an N-alkylated tranylcypromine derivative and potent lysine-specific demethylase 1 (LSD1) inhibitor used in epigenetic research. It is supplied as a solid or as a DMSO solution for biochemical and cellular studies; refer to the certificate of analysis for batch-specific purity and handling information.
  • Potent inhibitor of lysine-specific demethylase 1 (LSD1).
  • High purity, 98.1% by HPLC.
  • Available as solid powder and 10 mM solution in DMSO.
  • Chemical formula C22H26ClF2N3O2 and molecular weight 437.91 g/mol.
  • Intended for research use only; not for human use.