Learn More
Medchemexpress LLC Ethanone, 1-[4-[3-[4-(1H-benzimidazol-2-ylcarbonyl)phenoxy]-2-pyrazinyl]-1-piperidinyl]- | 1227067-61-9 | 99.0% | 25 MG
SDP
Catalog No. 5000342724
Encompass_Preferred
AMG 579 is a potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A) with an IC50 of 0.1 nM. This product is for research use only.
  • Potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A).
  • IC50 of 0.1 nM for phosphodiesterase 10A.
  • Shows statistically significant reduction of PCP induced behavior in rats.
  • Exhibits superior oral bioavailability of 72% in dogs.
Catalog No. 50-003-42724 Supplier Medchemexpress LLC Supplier No. 25MGHY12913
AMG 579 is a potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A) with an IC50 of 0.1 nM. This product is for research use only.
  • Potent, selective, and efficacious inhibitor of phosphodiesterase 10A (PDE10A).
  • IC50 of 0.1 nM for phosphodiesterase 10A.
  • Shows statistically significant reduction of PCP induced behavior in rats.
  • Exhibits superior oral bioavailability of 72% in dogs.