Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1) | 37845-71-9 | 98.3% | 311.64 | 50 MG
SDP

Catalog No. 5000403638
Encompass_Preferred

Clenbuterol impurity 1 (HY-33450) is a white to off-white solid with a molecular weight of 311.64 and a purity of 98.26% by HPLC. It is identified by CAS number 37845-71-9 and has the chemical name Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1).

  • Purity of 98.26%
  • White to off-white solid
  • Molecular formula C12H17Cl3N2O
  • Stable at room temperature for 3 years
  • Stable in solvent for 2 years at -80°C

Catalog No. 50-004-03638 Supplier Medchemexpress LLC Supplier No. HY3345050MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clenbuterol impurity 1 (HY-33450) is a white to off-white solid with a molecular weight of 311.64 and a purity of 98.26% by HPLC. It is identified by CAS number 37845-71-9 and has the chemical name Ethanone, 1-(4-amino-3,5-dichlorophenyl)-2-[(1,1-dimethylethyl)amino]-, hydrochloride (1:1).

  • Purity of 98.26%
  • White to off-white solid
  • Molecular formula C12H17Cl3N2O
  • Stable at room temperature for 3 years
  • Stable in solvent for 2 years at -80°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.