Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ethanone, 1-(6-methyl-3-pyridinyl)-2-[4-(methylthio)phenyl]- | 221615-72-1 | ≥98.0% | C15H15NOS | 250 MG
SDP

Catalog No. 5000358437
Encompass_Preferred

Pantoprazole impurity 43 is an impurity of Pantoprazole. This product is for research use only and not for sale to patients. It is a controlled substance and may not be for sale in all territories.

  • Appearance: Solid
  • Molecular weight: 257.35
  • Purity: ≥98.0%

Catalog No. 50-003-58437 Supplier Medchemexpress LLC Supplier No. 250MGHY59363
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pantoprazole impurity 43 is an impurity of Pantoprazole. This product is for research use only and not for sale to patients. It is a controlled substance and may not be for sale in all territories.

  • Appearance: Solid
  • Molecular weight: 257.35
  • Purity: ≥98.0%

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.