Learn More
Medchemexpress LLC Ethanone, 2-bromo-1-(4-methoxyphenyl)- | 2632-13-5 | 98.9% | 229.07 | 25 G
SDP
Catalog No. 5000453972
Encompass_Preferred
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor that exhibits anti-angiogenic effects. It is intended for research use only.
  • Purity: 99.55%
  • Anti-angiogenic effect: Exhibits anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.
Catalog No. 50-004-53972 Supplier Medchemexpress LLC Supplier No. 25GHYW013478
PTP inhibitor 1 is a protein tyrosine phosphatase (PTP) inhibitor that exhibits anti-angiogenic effects. It is intended for research use only.
  • Purity: 99.55%
  • Anti-angiogenic effect: Exhibits anti-angiogenic activity with an IC50 of 3.7 μM for HUVECs.