Learn More
Medchemexpress LLC Ethanone, 2-hydroxy-1-phenyl- | 582-24-1 | 136.15 | 10 G
SDP
Catalog No. 5000447844
Encompass_Preferred
2-Hydroxyacetophenone is a principal root volatile of Carissa edulis that can be used as a building block. It shows inhibitory effects on infection of HIV/SARS-CoV S pseudovirus with an IC50 of 1.8 mM and is also a substrate for Lactobacillus brevis alcohol dehydrogenase (LBADH).
  • Inhibits HIV/SARS-CoV
  • Principal root volatile of Carissa edulis
  • Can be used as a building block
  • Substrate for Lactobacillus brevis alcohol dehydrogenase (LBADH)
Catalog No. 50-004-47844 Supplier Medchemexpress LLC Supplier No. HYW00219810G
Add to Cart
Add to Cart
2-Hydroxyacetophenone is a principal root volatile of Carissa edulis that can be used as a building block. It shows inhibitory effects on infection of HIV/SARS-CoV S pseudovirus with an IC50 of 1.8 mM and is also a substrate for Lactobacillus brevis alcohol dehydrogenase (LBADH).
  • Inhibits HIV/SARS-CoV
  • Principal root volatile of Carissa edulis
  • Can be used as a building block
  • Substrate for Lactobacillus brevis alcohol dehydrogenase (LBADH)