Learn More
Medchemexpress LLC Ethanone, 2-methoxy-1,2-diphenyl | 3524-62-7 | 98.1% | 226.27 g·mol⁻¹ | C15H14O2 | 1 G
SDP
Supplier:  Medchemexpress LLC HY1185711G
Encompass_Preferred
Benzoin methyl ether is an organic photoinitiator (CAS 3524-62-7) provided for research use. It is a white to off-white solid with a molecular weight of 226.27 g·mol⁻¹ and is supplied at high purity for photochemistry and polymerization experiments.
  • used as a photoinitiator in laboratory photochemistry and polymerization research.
  • high purity suitable for analytical and preparative work.
  • white to off-white solid, easy to weigh and handle.
  • molecular formula C15H14O2; molecular weight 226.27 g·mol⁻¹.
  • store refrigerated and protected from air to preserve stability.
  • available in small laboratory pack sizes for research workflows.
Catalog No. 50-002-26294