Learn More
AdipoGen Ethanoyl-DL-homoserine lactone
SDP
Supplier:  AdipoGen CDXE0071M005
Encompass_Preferred
Chemical. CAS 114375-78-9. Formula C6H9NO3. MW 143.14. Synthetic N-Ethanoyl-DL-homoserine lactone is the shortest alkyl homologue and most polar of a family of mediators of cell to cell interactions in bacterial biofilms. Acylhomoserine lactones have been detected in hundreds of bacterial species. N-acylhomoserine lactones AHL are involved in quorum sensing, controlling gene expression, and cellular metabolism. N-Ethanoyl-DL-homoserine lactone is not found in nature and can serve as a polar negative control for quorum sensing events.
Catalog No. 50-256-0976