Learn More
Medchemexpress LLC EthD bromide | 1239-45-8 | 99.27% | 250 MG
SDP
Catalog No. 5000413747
Encompass_Preferred
EthD bromide is an intercalating agent commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis. It binds to DNA for visualization under UV light.
  • Fluorescent tag for nucleic acids
  • Intercalating agent
  • Used for agarose gel electrophoresis
  • Used in studies to reduce mitochondrial DNA copy number
  • Used in Western blot analysis of proteins in the cGAS-STING pathway
  • Visualizes EtBr uptake via hemichannels in astrocytes
  • Targets DNA for staining
  • Involves the cell cycle/DNA damage pathway
Catalog No. 50-004-13747 Supplier Medchemexpress LLC Supplier No. 250MGHYD0021
EthD bromide is an intercalating agent commonly used as a fluorescent tag (nucleic acid stain) in molecular biology laboratories for techniques such as agarose gel electrophoresis. It binds to DNA for visualization under UV light.
  • Fluorescent tag for nucleic acids
  • Intercalating agent
  • Used for agarose gel electrophoresis
  • Used in studies to reduce mitochondrial DNA copy number
  • Used in Western blot analysis of proteins in the cGAS-STING pathway
  • Visualizes EtBr uptake via hemichannels in astrocytes
  • Targets DNA for staining
  • Involves the cell cycle/DNA damage pathway